missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,1'-Bi(2-naphthylamine), 97%, Thermo Scientific Chemicals

Catalog Number p-7028459
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
18531-95-8
C20H16N2
284.362
MFCD00145204
DDAPSNKEOHDLKB-UHFFFAOYSA-N
1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
20571
1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)N)N
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
18531-95-8
C20H16N2
284.362
MFCD00145204
DDAPSNKEOHDLKB-UHFFFAOYSA-N
1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
20571
1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)N)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico (S)-(-)-1,1'-Bi(2-naphthylamine)
Punto di fusione 236°C to 242°C
Quantità 250 mg
Beilstein 2419071
Optical Rotation −150° (c=1 in Pyridine)
Peso formulazione 284.36
Percent Purity 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'