missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-1-Boc-3-aminopyrrolidine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantità:
1 g
5 g
Descrizione
(S)-(-)-1-Boc-3-aminopyrrolidine is specifically used to protect amine in the solid phase synthesis of peptides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(-)-1-Boc-3-aminopyrrolidine is specifically used to protect amine in the solid phase synthesis of peptides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
(S)-(-)-1-Boc-3-aminopyrrolidine is specifically used to protect amine in the solid phase synthesis of peptides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifica
Materiale o nome chimico | (S)-(-)-1-Boc-3-aminopyrrolidine |
Punti di ebollizione | 115°C (4 mmHg) |
Note percentuale saggio | ee 99% |
Quantità | 1 g |
Numero UN | UN2735 |
Beilstein | 8312125 |
Optical Rotation | −3° (Neat) |
IUPAC Name | tert-butyl 3-aminopyrrolidine-1-carboxylate |
Peso formulazione | 186.25 |
Percent Purity | 98% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'