Learn More
(S)-(+)-4-Phenyl-2-oxazolidinone, 99%, Thermo Scientific Chemicals
Catalog Number p-183252
Click to view available options
Quantità:
1 g
5 g
Imballaggio:
Glass Bottle
99395-88-7
C9H9NO2
163.18
MFCD00043396
QDMNNMIOWVJVLY-MRVPVSSYSA-N
s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076
730424
(4S)-4-phenyl-1,3-oxazolidin-2-one
C1C(NC(=O)O1)C2=CC=CC=C2
99395-88-7
C9H9NO2
163.18
MFCD00043396
QDMNNMIOWVJVLY-MRVPVSSYSA-N
s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076
730424
(4S)-4-phenyl-1,3-oxazolidin-2-one
C1C(NC(=O)O1)C2=CC=CC=C2
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico (S)-(+)-4-Phenyl-2-oxazolidinone
Punto di fusione 130°C to 133°C
Nota nome 99%
CAS min. % 98.5
CAS max % 100.0
Colore White
Intervallo percentuale saggio 98.5% min. (GC)
Spettro a infrarossi Authentic
Imballaggio Glass Bottle
Rotazione specifica + 49.5
Peso formulazione 163.18
Percent Purity 99%
Condizione di rotazione specifica +49.5° (20°C c=2,CHCl3)
Quantità 1 g
Forma fisica Crystalline Powder