Learn More
Heidolph Instruments S-Box Package 1
Product Code. 17878000 Shop All Heidolph Instruments Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Violet
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17878000 Violet Each
17848000 Green Each
17858000 Orange Each
17868000 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17878000 Supplier Heidolph Instruments Supplier No. 4000500300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Este artículo no se puede devolver. Vea la política de devoluciones
The S-Box Package 1 includes additional accessories to complete an existing S-Biosystem incubation system.
  • Each S-box is a stand-alone miniature incubator.
  • Inside the S-box there is room for one standard multi-well plate, or one cell culture bottle (up to T75).
  • The complete interior (inner box) can be purchased as a sterilized exchangeable element in a sterile outer packaging.
  • Your culture is prevented from drying out by a sachet of sterilized water (Waterbag), which is enough for weeks.
  • To avoid mix-up, the front cover of the S-box is available in different colors.
TRUSTED_SUSTAINABILITY
Especificaciones
Color Violet
Includes 1 S-box, 1 Tool Kit, 4 inner boxes, 4 water bags, 4 front covers
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.