missing translation for 'onlineSavingsMsg'
Learn More

(S)-alpha-Methylaspartic acid, 98%, 98% ee, Thermo Scientific™

Catalog Number p-56524
Click to view available options
Quantità:
100 mg
500 mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
3227-17-6
C5H8NO4
146.12
MFCD00145254
CWAYDJFPMMUKOI-YFKPBYRVSA-M
alpha-methyl-l-asp, 2-methyl-l-aspartic acid, s-+-2-amino-2-methylbutanedioic acid, 2s-2-amino-2-methylbutanedioic acid, alpha-methylaspartic acid, aspartic acid, 2-methyl, ambotzhaa7550, a-methyl-l-asp, alpha-methyl aspartic acid, h-a-me-asp-oh
2724884
C[C@]([NH3+])(CC([O-])=O)C([O-])=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
3227-17-6
C5H8NO4
146.12
MFCD00145254
CWAYDJFPMMUKOI-YFKPBYRVSA-M
alpha-methyl-l-asp, 2-methyl-l-aspartic acid, s-+-2-amino-2-methylbutanedioic acid, 2s-2-amino-2-methylbutanedioic acid, alpha-methylaspartic acid, aspartic acid, 2-methyl, ambotzhaa7550, a-methyl-l-asp, alpha-methyl aspartic acid, h-a-me-asp-oh
2724884
C[C@]([NH3+])(CC([O-])=O)C([O-])=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico (S)-α-Methylaspartic acid
Nota nome 99%
CAS min. % 97.5
CAS max % 100.0
Colore White
Eccesso enantiomerico 98.0% min. (HPLC)
Spettro a infrarossi Authentic
Intervallo percentuale saggio 98%
Formula lineare HOOCC(CH3)nH2CH2COOH
Quantità 100 mg
Rotazione specifica +46° to +54° (20°C, 589nm) (c=0.14 in water)
IUPAC Name (2S)-2-azaniumyl-2-methylbutanedioate
Peso formulazione 147.13
Percent Purity ≥97.5%
Forma fisica Fine Crystalline Powder or Powder
Vedi altri risultati Mostra meno risultati
Identificativo prodotto
  • (S)-alpha-Methylaspartic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'