Learn More
(S)-Salsoline Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | (S)-Salsoline Hydrochloride |
| CAS | 881-26-5 |
| Formula molecolare | C11 H15 N O2 • HCl |
| Quantità | 2.5 mg |
| Sinonimo | (1S)-1,2,3,4-Tetrahydro-7-methoxy-1-methyl-6-isoquinolinol Hydrochloride, (-)-Salsoline, (1S)-1,2,3,4-Tetrahydro-7-methoxy-1-methyl-6-Isoquinolinol Hydrochloride, (1S)-7-Methoxy-1-methyl-1,2,3,4-tetrahydroisoquinolin-6-ol Hydrochloride, (S)-Salsoline Hydrochloride, 7-O-Methylsalsolinol Hydrochloride, Methoxysalsolinol Hydrochloride, O7-Methylsalsolinol Hydrochloride |
| InChI Formula | InChI=1S/C11H15NO2.ClH/c1-7-9-6-11(14-2)10(13)5-8(9)3-4-12-7;/h5-7,12-13H,3-4H2,1-2H3;1H/t7-;/m0./s1 |
| SMILES | C[C@H]1C2=CC(OC)=C(O)C=C2CCN1.Cl |
| IUPAC Name | (S)-7-methoxy-1-methyl-1,2,3,4-tetrahydroisoquinolin-6-ol hydrochloride |
| Molecular Weight (g/mol) | 229.7 |
| Peso formulazione | 229.09 |