missing translation for 'onlineSavingsMsg'
Learn More

S100A5 Antibody (S100A5/7475), Novus Biologicals™

Codice prodotto. 18630535 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
100 μg
20 μg
Dimensione della confezione:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18630535 20 μg 20µg
18664015 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18630535 Fornitore Novus Biologicals N. del fornitore NBP32384820ug

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

S100A5 Monoclonal antibody specifically detects S100A5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigene S100A5
Applicazioni Immunohistochemistry (Paraffin)
Classificazione Monoclonal
Clone S100A5/7475
Concentrazione 0.2 mg/mL
Coniugato Unconjugated
Diluizione Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulazione 10 mM PBS with 0.05% BSA
Alias gene Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5
Specie ospite Mouse
Immunogeno Recombinant fragment (around aa1-92) of human S100A5 protein (exact sequence is proprietary)
Metodo di purificazione Protein A or G purified
Quantità 20 μg
Status giuridico RUO
Disciplina di ricerca Neuroscience, Signal Transduction
Primario o secondario Primary
ID gene (immissione) 6276
Target Species Human
Content And Storage Store at 4°C.
Forma Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.