Learn More
Hirschmann™ Safe-Cone Filters for Labopette™
Codice prodotto. 10012223
Cambia vista
Click to view available options
Volume (sistema metrico):
Var: 10-100/20-200 μL, Fixed: 100/200 μL
Var: 10-500/50-1,000/500-5,000/50-1200 μL, 100-1000 μL, Fixed: 250/500/1000 μL
Var: 1000-5000 μL, Fixed: 2000/5000 μL
Var: 2-20/5-50 μL, Fixed 20/25/50 μL
Dimensione della confezione:
pezzi 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Volume (sistema metrico) Dimensione della confezione
10012223 Var: 2-20/5-50 μL, Fixed 20/25/50 μL pezzi 50
10233151 Var: 1000-5000 μL, Fixed: 2000/5000 μL pezzi 50
10052173 Var: 10-100/20-200 μL, Fixed: 100/200 μL pezzi 50
10173913 Var: 10-500/50-1,000/500-5,000/50-1200 μL, 100-1000 μL, Fixed: 250/500/1000 μL pezzi 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 10012223 Fornitore Hirschmann™ N. del fornitore 9470013
  • ONLINE EXCLUSIVE PRICE
    80.12€
    58.90€ / pezzi 50
    Risparmia 21.22€
    26% Sconto
In stock
For the prevention of cross contamination of liquid samples or vapours when pipetting radioactive, biohazardous or corrosive samples.
  • Provides the decisive advantage when protecting pipettes and samples against contamination
  • Extends the service life of pipettes
  • Provides excellent protection for the internal components if replaced regularly
  • Can serve as additional protection against penetration of liquids into the interior of the pipette
  • For use with a range of Labopette™ models
Specifica
Tipo Safety Filter
Volume (sistema metrico) Var: 2-20/5-50 μL, Fixed 20/25/50 μL
Da utilizzare con (applicazione) Protection against cross-contamination
Da utilizzare con (apparecchiatura) Labopette models 9475406, 9475407, 9475503, 9475504, 9475505, 9475506, 9475602, 9475702