Learn More
Ansell Edmont™ Safe-Knit™ GP 72-265 Series Grey Polyester/Glass Fiber Heavy Weight Gloves
Sfoglia Tutto Ansell ProdottiDescription
- Safe-Knit™ GP sleeves contain a higher proportion of Dyneema™ fibers which are as strong as steel fiber but much lighter and more flexible.
- Safe-Knit™ GP Sleeves provide optimal protection from cuts and abrasions, while their light weight and comfortable fit ensure high levels of user acceptance.
Specifications
Specifications
| Materiale parte posteriore | Polyester/Glass Fiber |
| Certificazione/Conformità | Standard CATEGORY II EN 388 : 354X |
| Materiale polso | Polyester/Glass Fiber |
| Product Type | Heavyweight Gloves |
| Stile polso guanto | Attached Sleeve |
| Materiale rivestimento interno | Dyneema™ |
| Rivestimento interno | Dyneema™ |
| Quantità | 72 Pairs |
| Resistente a | Cut resistant |