Learn More
3M™ Safety-Walk Edge Sealing Compound
Product Code. 17267610
Change view
Click to view available options
Quantity:
150 mL
Unit Size:
12 bottles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17267610 150 mL 12 bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17267610 Supplier 3M™ Supplier No. 7100244310
  • ONLINE EXCLUSIVE PRICE
    574.00€
    331.07€ / pezzi 12
    Risparmia 242.93€
    42% Sconto
 Restricted Item
Spedizione stimata: 11-12-2026
Liquid, solvent-based sealer that protects exposed edges of slip-resistant tapes and treads from excessive moisture or liquids.
  • Protect the exposed edges of slip-resistant tapes and treads
  • Keep surfaces safe from excessive moisture and liquids
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Quantity 150 mL
Volume (Metric) 150 mL