Descrizione
SCAT™ Safety Caps V2.0 provide maximum safety for your HPLC solvent supply.
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of connections
- Improved design for easy connection of capillaries in various colors
- Free air valve with change indicator included with every Safety Cap
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Lab Bottle |
| Include | 1 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve |
| Materiale | PTFE |
| Dimensioni filo | GL/S40 |
| Da utilizzare con (applicazione) | HPLC Systems |
| Product Type | Safety Cap |