Learn More
HALYARD™ SAFEVIEW™ Protective Eyewear, Assembled Glasses
Codice prodotto. 30274753 Sfoglia Tutto Halyard Prodotti
Cambia vista
Click to view available options
Quantità:
50
Dimensione della confezione:
pezzi 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
30274753 50 pezzi 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30274753 Fornitore HALYARD™ N. del fornitore SV50A
Spedizione stimata: 11-12-2026
SAFEVIEW™ Eyewear protects eyes in splashing or spraying environments. Lightweight and latex-free, these glasses comfortably fit over most prescription eyewear and come in various colorful frames.
  • HALYARD™ SAFEVIEW™ Eyewear provides reliable eye protection for healthcare professionals and other users in environments where splashing or spraying may occur.
  • For use in situations which may involve splashing or spraying
  • Certified as PPE Category II, for moderate risks
  • Lightweight design to reduce strain and increase comfort in use
  • Colorful frames appeal to a variety of preferences
  • Designed to be easily worn over most prescription glasses
  • Constructed from natural rubber latex-free and DEHP-free materials
  • Lenses film: clear polyester; Frames: polypropylene
Specifica
Certificazione/Conformità PPE Regulation 2016/425 (PPE category II)
Lens Coating Uncoated
Lens Tint Clear
Product Type Over-the-Glasses Protective Eyewear
Colore telaio Assorted colours
N. per custodia 50
Quantità 50
N. per confezione 10
Size One-size
Monouso Limited-use
Lens Material Polyester
UV Protection None
Frame Type Standard