Descrizione
- Disposable hygiene labcoats protect the work environment from particles coming from the operator's body (skin flakes, hair, sweat, etc.)
- They are suitable for HACCP procedures
- They help the operator protect themselves from the risk of stains related to their activity
Specifica
Specifica
| Size | Small |
| Sesso | Unisex |
| Materiale | Polypropylene |
| Colore | White |
| Tipo di chiusura | Zipper |
| Stile polso | Elastic |
| Monouso | Single-use |
| Sterilità | Non-sterile |
| Resistenza | None |
| Stile | Knee-length |
| Vedi altri risultati |