Learn More
Salicylic Acid Methyl Ether-d3, TRC
Specifica
Specifica
| Materiale o nome chimico | Salicylic Acid Methyl Ether-d3 |
| CAS | 96739-36-5 |
| Formula molecolare | C8H5D3O3 |
| Quantità | 250 mg |
| Sinonimo | o-Methoxybenzoic Acid-d3, o-Anisic Acid-d3 (8CI), 2-Methoxybenzoic Acid-d3, NSC 3778-d3, O-Methylsalicylic Acid-d3 |
| InChI Formula | InChI=1S/C8H8O3/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5H,1H3,(H,9,10)/i1D3 |
| SMILES | [2H]C([2H])([2H])OC1=C(C(O)=O)C=CC=C1 |
| IUPAC Name | 2-(methoxy-d3)benzoic acid |
| Molecular Weight (g/mol) | 155.17 |
| Peso formulazione | 155.07 |