Learn More
Cole-Parmer™ Salt Windows
Product Code. 11765349 Shop All Cole-Parmer Products
Change view
Click to view available options
For Use With (Application):
IR Spectroscopy
Thickness:
4 mm
Diameter (Metric):
25mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Thickness Diameter (Metric) Unit Size
11765349 IR Spectroscopy 4 mm 25mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11765349 Supplier Cole-Parmer™ Supplier No. 8300156
  • ONLINE EXCLUSIVE PRICE
    90.00€
    59.30€ / pezzi 1
    Risparmia 30.70€
    34% Sconto
Spedizione stimata: 25-01-2027
Cole-Parmer Salt Windows, For Use With: IR Spectroscopy, Quantity: 1Each, Sterile, Thickness: 4mm
Specifications
Type Salt Window
Diameter (Metric) 25mm
For Use With (Application) IR Spectroscopy
Quantity 1 Ea.
Thickness 4 mm