Learn More
S Murray and Co™ SAMCO™ Stainless Steel Scissors
Product Code. 12912055
Change view
Click to view available options
Length (Metric):
115 mm
125 mm
Blade Style:
Straight
Tip Style:
Fine Point
Very Fine Point
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Blade Style Tip Style Unit Size
12912055 125 mm Straight Fine Point Each
12348099 115 mm Straight Very Fine Point Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12912055 Supplier S Murray and Co™ Supplier No. E803/02
  • ONLINE EXCLUSIVE PRICE
    3.10€
    2.62€ / pezzi 1
    Risparmia 0.48€
    15% Sconto
Spedizione stimata: 15-12-2026
Use these scissors for a wide range of applications. S Murray™ SAMCO™ Stainless Steel Scissors are available in a variety of blade and tip styles to meet your needs.
  • Fully guaranteed in normal use and subject to stringent quality inspection procedures
Specifications
Product Type Stainless Steel Scissors
Blade Style Straight
Length (Metric) 125 mm
Material Stainless Steel
Description Fine points and closed shanks
Tip Style Fine Point