Learn More
Thermo Scientific™ Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Thermo Scientific™ sample and skimmer cones for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova series ICP-MS systems are available to suit your analysis requirements and designed for durability, performance and ease of use.
Assure robust, uncomplicated trace elemental analysis for your laboratory with the Thermo Scientific iCAP MX and iCAP Q/Qnova series ICP-MS systems. Our easily changeable Ni and Pt sample and skimmer cones are available to suit your analysis requirements and designed for durability, performance and ease of use.
Select high-quality consumables for consistent performance. These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in either solid Ni construction or Pt tipped for even greater durability and specific analysis requirements.
- Sample cones made from nickel and platinum
- Skimmer cones made from nickel and platinum
- Platinum, boron free skimmer cone (for semiconductor-related applications)
- Skimmer cone inserts with different channel length to tailor instrument characteristics for each application
Specifica
Specifica
| Tipo | Skimmer Cone Insert 4,5 (Robust Interface) |
| Da utilizzare con (apparecchiatura) | iCAP Q/Qnova Series ICP-MS |
| Include | Skimmer Cone Insert 4.5 Robust |