Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S protein RBD (Y453F), His Tag (MALS verified)
Marca:  ACROBiosystems SPD-C52Hk-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17226520
  • dietro preventivo / 100µg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
QHD43416.1 (Y453F)
>95% as determined by SDS-PAGE.
SARS-CoV-2 S protein RBD (Y453F), His Tag (SPD-C52Hk) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (Y453F)).
-20°C
S protein RBD
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
SARS-CoV-2
26.5 kDa
100 µg
Arg 319 - Lys 537
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
Unconjugated
PBS, pH7.4
HEK293
Viral Proteins