Learn More
ACROBiosystems SARS-CoV-2 S protein (F817P, A892P, A899P, A942P, K986P, V987P), His Tag (MALS verified)
Marca: ACROBiosystems SPN-C52Hd-50ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 138.2 kDa. The protein migrates as 75-85 kDa, 90-100 kDa and 150-160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
QHD43416.1 | |
>95% as determined by SDS-PAGE. | |
SARS-CoV-2 S protein, His Tag (SPN-C52Hd) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein. | |
-20°C | |
S protein | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
SARS-CoV-2 |
138.2 kDa | |
50 µg | |
Val 16 - Pro 1213 | |
Spike,S protein,Spike glycoprotein,S glycoprotein,COVID-19 | |
Unconjugated | |
PBS | |
HEK293 | |
Viral Proteins |