missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems SARS-CoV-2 S protein (F817P, A892P, A899P, A942P, K986P, V987P), His Tag (MALS verified)
587.89€ - 4062.99€
Specifica
Numero di accesso | QHD43416.1 |
---|---|
Peso molecolare | 138.2 kDa |
Sorgente | SARS-CoV-2 S protein, His Tag (SPN-C52Hd) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein. |
Immunogeno | Val 16 - Pro 1213 |
Requisiti di stoccaggio | -20°C |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17286550
|
ACROBiosystems
SPN-C52HD-500UG |
500 μg |
4062.99€
500µg |
Spedizione stimata: 02-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
17296550
|
ACROBiosystems
SPN-C52HD-50UG |
50 μg |
587.89€
50µg |
Spedizione stimata: 02-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 138.2 kDa. The protein migrates as 75-85 kDa, 90-100 kDa and 150-160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
QHD43416.1 | |
SARS-CoV-2 S protein, His Tag (SPN-C52Hd) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein. | |
-20°C | |
S protein | |
SARS-CoV-2 | |
PBS | |
HEK293 | |
Viral Proteins |
138.2 kDa | |
Val 16 - Pro 1213 | |
Spike,S protein,Spike glycoprotein,S glycoprotein,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE. |