missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems SARS S protein (R667A, K968P, V969P), His Tag (MALS verified)

699.64€ - 5610.07€

Specifica

Numero di accesso AAP13567.1 (R667A, K968P, V969P)
Peso molecolare 136.3 kDa
Sorgente SARS S protein (R667A, K968P, V969P), His Tag (SPN-S52H6) is expressed from human 293 cells (HEK293). It contains AA Ser 14 - Pro 1195 (Accession # AAP13567.1 (R667A, K968P, V969P)). The recombinant protein is expressed with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (K968P, V969P) and alanine substitutions (R667A) are introduced to stabilize the trimeric prefusion state of SARS-CoV S protein and abolish the furin cleavage site, respectively.)
Immunogeno Ser 14 - Pro 1195
Requisiti di stoccaggio -20°C
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
17226570
Vedere i documenti Dettagli della promozione
ACROBiosystems
SPN-S52H6-100UG
100 μg
699.64€
100µg
Visualizzare prodotti alternativi
Risparmia fino a 
»
Spedizione stimata: 10-04-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
17236570
Vedere i documenti Dettagli della promozione
ACROBiosystems
SPN-S52H6-1MG
1 mg
5610.07€
1mg
Visualizzare prodotti alternativi
Risparmia fino a 
»
Spedizione stimata: 10-04-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descrizione

Descrizione

It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 136.3 kDa. The protein migrates as 190-210 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica

Specifica

AAP13567.1 (R667A, K968P, V969P)
SARS S protein (R667A, K968P, V969P), His Tag (SPN-S52H6) is expressed from human 293 cells (HEK293). It contains AA Ser 14 - Pro 1195 (Accession # AAP13567.1 (R667A, K968P, V969P)). The recombinant protein is expressed with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (K968P, V969P) and alanine substitutions (R667A) are introduced to stabilize the trimeric prefusion state of SARS-CoV S protein and abolish the furin cleavage site, respectively.)
-20°C
S protein
SARS
PBS
HEK293
Viral Proteins
136.3 kDa
Ser 14 - Pro 1195
Spike,S protein,Spike glycoprotein,S glycoprotein,COVID-19
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE.
Videos
SDS
Documenti

Documenti

Product Certifications