missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Sartopore™ 2 MidiCaps Polypropylene Housing Polyethersulfone Membrane Filter Capsules
Marca: Sartorius™ 5445307H8G-OO--A
Dettagli aggiuntivi : Peso : 0.10000kg
Specifica
Autoclavable | |
PES | |
Sterile | |
For therapeutics, biological fluids, injectables, media, buffers, chemicals, cleaning and sanitizing agents filtering applications | |
0.5 in. Hose Barb | |
0.5 in. Hose Barb | |
Filter Capsule | |
77 mm |
ISO 9001, USP Class VI, cGMP Standard, HIMA and ASTM F-838-05 | |
0.2 μm | |
0.1 m2 | |
Polypropylene | |
4Pack | |
4Pack | |
148 mm |