Learn More
Sartorius Funnels for Combisart™
Product Code. 10446741 Shop All Sartorius Products
Change view
Click to view available options
Product Type:
Funnel
Funnel Lid
Includes:
Locking Clamp
Small Central Air Port
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Includes Unit Size
10446741 Funnel Locking Clamp Each
10243541 Funnel Lid Small Central Air Port Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10446741 Supplier Sartorius Supplier No. 6981002
  • ONLINE EXCLUSIVE PRICE
    397.00€
    366.33€ / pezzi 1
    Risparmia 30.67€
    7% Sconto
Spedizione stimata: 16-12-2026
Reusable stainless steel Combisart™ Funnels for microbial enumeration and particle counting.
  • Specially designed for use in microbiological quality assurance, these three filter holders differ only in their capacity
  • The reusable funnels feature a special locking clamp that simplifies the positioning and removal of the membrane filter, ensuring secure lock-down
  • For traceability, each filter funnel has an individual serial number
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Material Stainless Steel
For Use With (Equipment) Combisart Funnel
Includes Locking Clamp
Quantity 1 Ea.
Product Type Funnel