Learn More
Sartorius Quantitative Grade 1289 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Marca:  Sartorius FT-3-207-240
Dettagli aggiuntivi : Peso : 0.10000kg
Codice prodotto. 10778772
  • 65.50€ / pezzi 100
Spedizione stimata: 20-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifica
Specifica
Disc
20 sec.
100/Pk.
240 mm
8 to 12 μm
1289
0.21 mm