Learn More
Beskrivelse
Sartolab RF/BT are disposable vacuum filtration units. The units including receiver flask (RF) contain an optimized Sartorius PES membrane and combine highest flow-rates and throughput with extremely low protein binding and extractables. They are therefore the product of choice for sterile filtration of tissue culture media, buffers, additives and other aqueous biologicals. Sartolab RF vacuum units are available as 150, 250, 500 and 1000mL units. Low protein binding Fast flow and high throughput. Easy to use. Includes receiver flask.
Tekniske data
Tekniske data
| Connettori | Plug-on hose nipple |
| Monouso | Single-use |
| Sterilità | Sterile |
| Area di filtraggio | 62 cm2 |
| Materiale alloggiamento | Polystyrene |
| Include | 150mL, 500mL, 1000mL funnel and receiver flask, 5cm and 9cm dia. 0.1μm pore size polyethersulfone membrane |
| Quantità | 12 Pack |
| Tipo | Sartolab RF Vacuum Filtration Unit |
| Volume (sistema metrico) | 1 L |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?