Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits
Codice prodotto. 15966181 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Descrizione:
SpeedVac SPD300DDA Vacuum Concentrator Kit
SpeedVac SPD300DDA Vacuum Concentrator Kit-No rotor
SpeedVac SPD300DDA Vacuum concentrator
Requisiti elettrici:
230 V 50 Hz
Dimensione della confezione:
Pezzo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Descrizione Requisiti elettrici Dimensione della confezione
15966181 SpeedVac SPD300DDA Vacuum Concentrator Kit 230 V 50 Hz Pezzo
15936181 SpeedVac SPD300DDA Vacuum concentrator 230 V 50 Hz Pezzo
15946181 SpeedVac SPD300DDA Vacuum Concentrator Kit-No rotor 230 V 50 Hz Pezzo
15956181 SpeedVac SPD300DDA Vacuum Concentrator Kit 230 V 50 Hz Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 15966181 Fornitore Thermo Scientific™ N. del fornitore SPD300P2230
  • ONLINE EXCLUSIVE PRICE
    68600.00€
    64200.00€ / pezzi 1
    Risparmia 4400.00€
    6% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 05-03-2027
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
  • For strong acids and bases used in combinatorial chemistry
TRUSTED_SUSTAINABILITY
Specifica
Certifications UL Listed to US and Canadian requirements. CE marked
Descrizione SpeedVac SPD300DDA Vacuum Concentrator Kit
Requisiti elettrici 230 V 50 Hz