Learn More
SB 203580 hydrochloride, Tocris Bioscience™
Selective inhibitor of p38 MAPK; water-soluble
Marca: Tocris Bioscience 1402/10
| Quantità | 10mg |
|---|
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Water-soluble salt of SB 203580 (Cat. No. 1202). Selective inhibitor of p38 mitogen-activated protein kinase (IC50 values are 50 and 500 nM for SAPK2a/p38 and SAPK2b/p38β2 respectively). Displays 100-500-fold selectivity over LCK, GSK3β and PKBα. Shown to inhibit interleukin-2-induced T cell proliferation, cyclooxygenase-1 and -2, and thromboxane synthase.Specifica
| SB 203580 hydrochloride | |
| C21H17ClFN3OS | |
| 4-[5-(4-Fluorophenyl)-2-[4-(methylsulphonyl)phenyl]-1H-imidazol-4-yl]pyridine hydrochloride | |
| 413.90 | |
| >98% |
| 869185-85-3 | |
| 10mg | |
| WOSGGXINSLMASH-UHFFFAOYNA-N | |
| 413.89 |