missing translation for 'onlineSavingsMsg'
Learn More

SCH772984, MedChemExpress

Codice prodotto. 16478108
Encompass_Preferred
Cambia vista
Click to view available options
Quantità:
5 mg
10 mg
50 mg
100 mg
200 mg
Dimensione della confezione:
100mg
10mg
200mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16478108 10 mg 10mg
16454027 50 mg -
16427876 100 mg 100mg
16411326 200 mg 200mg
16470568 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16478108 Fornitore Medchem Express N. del fornitore HY50846

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress SCH772984 is a highly selective and ATP-competitive ERK inhibitor, with IC50s of 4 and 1 nM for ERK1 and ERK2, respectively. SCH772984 has antitumor activity in MAPK inhibitor-naïve and MAPK inhibitor-resistant cells containing BRAF or RAS mutations.

SCH772984 is a highly selective and ATP-competitive ERK inhibitor, with IC50s of 4 and 1 nM for ERK1 and ERK2, respectively. SCH772984 has antitumor activity in MAPK inhibitor-naïve and MAPK inhibitor-resistant cells containing BRAF or RAS mutations.

Specifica

Materiale o nome chimico SCH772984
CAS 942183-80-4
Colore Yellow
Note sul grado di purezza Research
Formula molecolare C33H33N9O2
Quantità 10 mg
Informazioni di solubilità DMSO : 48 mg/mL (81.68 mM; Need ultrasonic)
SMILES O=C([C@H](CC1)CN1CC(N(CC2)CCN2C(C=C3)=CC=C3C4=NC=CC=N4)=O)NC5=CC6=C(C=C5)NN=C6C7=CC=NC=C7
Molecular Weight (g/mol) 587.67
Peso formulazione 587.67
Percent Purity 98.69%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) Cancer-Kinase/protease
Vedi altri risultati Mostra meno risultati

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.