Learn More
SI Analytics™ ScienceLine™ Single Electrode
Product Code. 10772893 Shop All SI Analytics Products
Change view
Click to view available options
Connector Types:
L 1 N Connection Cable
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
10772893 L 1 N Connection Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10772893 Supplier SI Analytics™ Supplier No. 1070046
  • ONLINE EXCLUSIVE PRICE
    201.00€
    183.00€ / pezzi 1
    Risparmia 18.00€
    8% Sconto
View all promotions
In stock
SI Analytics™ ScienceLine™ Single Electrode features a glass shaft and a platinum junction.
  • Ag/AgCl: KCl 3 mol/l, e.g. L 300
  • Reference electrode
TRUSTED_SUSTAINABILITY
Specifications
Connector Types L 1 N Connection Cable
Junction Platinum
Material Glass Shaft
pH Range 0 to 14
Type Reference Electrode
Reference Electrolyte Ag/AgCl
Temperature (Metric) Range -5°C to +100°C
Length (Metric) 120 mm
Diameter (Metric) Body 12 mm