missing translation for 'onlineSavingsMsg'
Learn More

SCP3 Mouse anti-Human, Clone: 1A11F1, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66409-1-IG-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898255

  • 121.38€ / 20µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregcy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.
Specifica

Specifica

SCP3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IZU3
SYCP3
SCP3 Fusion Protein Ag19330
20 μL
Primary
Human
Antibody
IgG1
Western Blot
1A11F1
Unconjugated
SYCP3
COR1, SCP 3, SCP3, SYCP3, synaptonemal complex protein 3
Mouse
Protein G
RUO
50511
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali