Learn More
sec-Butyl Acetate, TRC
Specifica
Specifica
| Materiale o nome chimico | Acetic acid-sec.-butyl ester |
| CAS | 105-46-4 |
| Formula molecolare | C6 H12 O2 |
| Quantità | 2.5 g |
| Sinonimo | Acetic acid, sec-butyl ester (8CI), sec-Butyl alcohol acetate (6CI), sec-Butyl alcohol, ester with AcOH (4CI), (±)-2-Butyl acetate, (±)-sec-Butyl acetate, 1-Methylpropyl acetate, 2-Acetoxybutane, 2-Butyl acetate, NSC 8034, Ultrasolve M 1200, sec-Butyl acetate, sec-Butyl ethanoate |
| InChI Formula | InChI=1S/C6H12O2/c1-4-5(2)8-6(3)7/h5H,4H2,1-3H3 |
| SMILES | CCC(C)OC(=O)C |
| IUPAC Name | butan-2-yl acetate |
| Molecular Weight (g/mol) | 116.16 |
| Peso formulazione | 116.0837 |