missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC14L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550751
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total SEC14L2 protein.
SEC14L2 Polyclonal antibody specifically detects SEC14L2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifica
SEC14L2 | |
Polyclonal | |
Unconjugated | |
SEC14L2 | |
1300013M05Rik; Alpha-tocopherol-associated protein; C22orf6; KIAA1186; KIAA1658; SEC14 like lipid binding 2; SEC14L2; SEC14-like 2; SEC14-like 2 (S. cerevisiae); SEC14-like lipid binding 2; SEC14-like protein 2; SEC14-like protein 2-like; SPF; squalene transfer protein; Supernatant protein factor; TAP; TAP1; tocopherol-associated protein; tocopherol-associated protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23541, 67815 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O76054, Q99J08 | |
SEC14L2 | |
Synthetic peptide corresponding to residues near the C terminal of human SEC14-like 2 (S. cerevisiae) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |