Learn More
Thermo Scientific™ Sensititre™ Companion Animal MIC Plate
Codice prodotto. 18850894 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Descrizione:
MIC Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Descrizione
18850894 MIC Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18850894 Fornitore Thermo Scientific™ N. del fornitore YCOMPAN1F
 Articolo soggetto a restrizioni
Spedizione stimata: 25-08-2026
Aggiungi al carrello
Aggiungi al carrello
Perform accurate AST with companion animal-specific antimicrobics, including cefovecin, with the Thermo Scientific™ Sensititre™ Companion Animal MIC Plate.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifica
Agenti antimicrobici Various
Descrizione MIC Plate
Codice antibiotico Various
Product Type MIC Plate
Test Type Minimum Inhibitory Concentration (MIC)
Isolati per piastra 1
Quantità 10/Pk.
Opzioni di lettura Autoread or manual