Learn More
Sensititre™ Gram Negative GNX3F AST Plate
Codice prodotto. 17779217
Cambia vista
Click to view available options
Quantità:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
17779217 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17779217 Fornitore Thermo Scientific Oxoid N. del fornitore YGNX3F
  • ONLINE EXCLUSIVE PRICE
    331.00€
    152.70€ / pezzi 10
    Risparmia 178.30€
    53% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 08-01-2027
Perform susceptibility testing of Gram-negative isolates with this research use only plate containing Colistin, Doripenem, Doxycycline and Polymyxin B
Perform susceptibility testing of non-fastidious Gram-negative isolates on a single platform with the Thermo Scientific™ Sensititre™ Gram Negative GNX3F AST Plate. The Sensititre™ Gram Negative GNX3F AST Plate is intended for research use only.
Decrease costs associated with multiple test methods with 21 antimicrobial compounds on a single plate, including Colistin and Polymyxin B.
Test most bacteria, including challenging resistant organisms, on a single platform with the Sensititre Gram Negative GNX3F AST Plate Plate.
  • Automated or manual read options are available for ultimate flexibility
  • Extended shelf life and room-temperature storage simplify inventory control
  • Individual packaging allows laboratories to test one plate at a time with no waste.
Not all products are available for sale in all territories. Please inquire. 
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifica
Standard McFarland 0,5 H2O (Part No. T3339)
Agenti antimicrobici Various
Tipo di brodo MHB (Part No. T3462)
Descrizione SensititreGram Negative Research Use Only MIC Plate for Non-Fermenters
Codice antibiotico Various
Product Type AST Plate
Target Organism Class Non-fastidious, Gram-negative Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Condizioni di incubazione 35°C O2
Tempo di incubazione 18 hr
Isolati per piastra 1
N. per confezione 10
Tipo di imballaggio Box
Quantità 10/Pk.
Opzioni di lettura Autoread or manual
Volume di trasferimento For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL
Volume per pozzetto 50 μL