Learn More
Thermo Scientific™ Sensititre™ Gram Positive MIC Plate
Codice prodotto. 17847144
Cambia vista
Click to view available options
Quantità:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
17847144 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17847144 Fornitore Thermo Scientific™ N. del fornitore YGPN3F
  • ONLINE EXCLUSIVE PRICE
    362.00€
    165.14€ / pezzi 1
    Risparmia 196.86€
    54% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 07-12-2026
Monitor resistance with a complete selection of antimicrobials using the Thermo Scientific™ Sensititre™ Gram Positive MIC Plate.
Monitor resistance with a complete selection of antimicrobials using the Sensititre Gram Positive MIC Plate.
  • Plate has fewest limitations for Enterococcus spp. and Staphylococcus spp. versus other automated systems
  • Full-range, true MIC readings provide results for better detection of resistance
  • Automated or manual read options are available for ultimate flexibility
  • MIC plate contains antimicrobic only; broth added at time of test extends shelf life and ensures accurate, easy-to-read results for Gram positive isolates
  • Extended 12 to 24 month shelf life and room-temperature storage simplify inventory control
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifica
Standard McFarland 0,5 H2O (Part No. T3339)
Agenti antimicrobici Various
Tipo di brodo MHB (Part No. T3462)
Descrizione Sensititre Gram Positive MIC Plate
Codice antibiotico Various
Product Type AST Plate
Target Organism Class Gram-positive Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Condizioni di incubazione 35°C O2
Tempo di incubazione 18-24 hr
Isolati per piastra 1
N. per confezione 10
Tipo di imballaggio Box
Quantità 10/Pk.
Opzioni di lettura Autoread or manual
Volume di trasferimento 10µL or 30µL* (*Aid in detection of resistance mechanisms for gram positive organisms; see pack insert)
Volume per pozzetto 50 μL