Descrizione
- Allergy-friendly certified by ECARF, the European Centre for Allergy Research Foundation.
- Recommended for more dirty hands that need more cleaning power, for example for use in kitchens.
- Provides extra care for sensitive skin and nurtures dry skin with lipid-replenishing ingredients like glycerine, betaine and panthenol.
- 96% of ingredients are from natural origin (according to specification and ISO16128, including water).
Specifica
Specifica
| Product Type | Hand Soap |
| Colore | White |
| Forma | Liquid |
| Imballaggio | Plastic Bottle |
| Quantità | 8/Pk. |
| Size | 525 mL |