Learn More
Septum for AcroSeal™, butyl rubber/teflon, size: 43 x 3 mm
Sfoglia Tutto Thermo Scientific Chemicals ProdottiDescrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Quantità | 10/Ea. |
| Size | 43 x 3mm |
| Materiale | Butyl Rubber, Teflon |