Descrizione
- Achieve easy and fast determination of melting and boiling points for professional quality control with the Melting Point system and accessories.
- Printer for GLP/GMP-compliant documentation of calibration results and melting and boiling point determination.
- Speed up input of sample parameters with the optional keyboard
- Ensure consistent sample preparation using compaction wire
Specifica
Specifica
| Da utilizzare con (applicazione) | Maintaining Documentation of Results |
| Da utilizzare con (apparecchiatura) | M-560/M-565 Melting Point Instrument |
| Modello | Star SP712 |