Learn More
BRANDTECH™ seripettor™ pro Pump Assembly
Product Code. 15643987 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15643987 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15643987 Supplier BRANDTECH™ Supplier No. 704551
  • ONLINE EXCLUSIVE PRICE
    141.00€
    126.91€ / pezzi 1
    Risparmia 14.09€
    9% Sconto
Spedizione stimata: 22-01-2027
For seripettor™ pro actuating unit
  • PPO
  • PEI (UV protection)
  • Hastaloy lifting spring
  • TRUSTED_SUSTAINABILITY
    Specifications
    Product Type Pump Assembly
    Volume (Metric) 2 mL