Learn More
Abeba™ Service 3000 Shoes
Descrizione
- CE
- Smooth leather
- Black
- Slip resistance outsole sole
- Sanitized treated
- Cushioned ankle supports
- Instep area with lacing
- Heel 45 mm
- Comfort foodbed
Specifica
Specifica
| Certificazione/Conformità | CE |
| Colore | Black |
| Materiale | Leather |
| Size | 41 |
| Caratteristiche suola | Slip-resistant |
| Stile | 3000 |