Learn More
Mettler Toledo™ SevenGo Duo™ Pro pH/Ion/Conductivity Multi Parameter Meters
Sfoglia Tutto Mettler Toledo ProdottiDescrizione
The instruments are suited for mobile measurements both in the field and in the production environment. The SG23 SevenGo Duo™ pH/mV/conductivity meter is best suited for parallel routine measurements in dual-channel mode. In addition, it also offers Intelligent Sensor Management (ISM™). The SG78 and SG68 SevenGo Duo Pro™ instruments allow simultaneous determination and display of two electrochemical parameters with highest possible accuracy. They have the same comprehensive GLP functions as their SevenGo Pro™ counterparts. In addition, the SevenGo Duo pro™ instruments have extra security functions, such as Intelligent Sensor Management (ISM ™), PIN-protected device logon and PIN-protected system setting.
- Portable instruments for pH, conductivity, ion and dissolved oxygen determination
- Robust, watertight single- and dual-channel instruments for use in demanding conditions
- Unique ergonomics and extremely simple operation
Specifica
Specifica
| Intervallo (conduttività) | 0.01 μS/cm to 1000 mS/cm |
| Intervallo (pH) | -2 to 20 |
| Intervallo (resistività) | 0.01 to 100.0 Mohm/cm |
| Intervallo (salinità) | 0 to 80 psu |
| Intervallo (TDS) | 0.1 mg/L to 600 g/L |
| Temperature Range (Metric) | -5°C to +130°C (pH ATC), -30°C to +130°C (MTC) |
| Include | Electrode Kit with InLab Expert Pro-ISM, InLab738-ISM (both IP67, 1.8m cable) |
| Modello | SG78-USP |
| Product Type | Electrode Kit |
| Intervallo (mV relativo) | -1999 to 1999 mV |