Learn More
SGE™ eVol™ Handheld Automated Analytical Syringe
Codice prodotto. 12907596
Cambia vista
Click to view available options
Volume (sistema metrico):
20 to 500 μL
5 μL
50 μL
500 μL
Stile del punto dell'ago:
Bevel
Without Needle
Quantità:
3/Pk.
Ea.
Dimensione della confezione:
Pezzo
pezzi 3
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Volume (sistema metrico) Stile del punto dell'ago Quantità Dimensione della confezione
12907596 20 to 500 μL Without Needle Ea. Pezzo
12927586 5 μL Bevel Ea. Pezzo
12997586 500 μL Bevel Ea. Pezzo
12947586 50 μL Bevel Ea. Pezzo
12927596 5 μL Bevel 3/Pk. pezzi 3
12937596 50 μL Bevel 3/Pk. pezzi 3
12957596 500 μL Bevel 3/Pk. pezzi 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Codice prodotto. 12907596 Fornitore SGE™ N. del fornitore 2910025
Spedizione stimata: 05-01-2027
Improves laboratory workflow
eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.
  • Digitally controlled positive displacement dispensing system
  • Programmable—enables user-independent liquid handling procedures
  • Easily calibrated to ensure accurate dispensing
  • Stores calibration factors for up to ten XCHANGE™ syringes
  • Touch Wheel user interface
  • Full color screen
Applications:
  • Preparation of calibration standards
  • Addition of internal standards
  • Precise dispensing of non-aqueous liquids
  • Sample dilution
Specifica
Product Type eVol™ Analytical Syringe, No Needle
Numero dei punti dell'ago No Needle Included
Quantità Ea.
Volume (sistema metrico) 20 to 500 μL
Stile del punto dell'ago Without Needle
Da utilizzare con (apparecchiatura) eVol Automated Syringe