Learn More
Trajan™ SilTite™ FingerTite Ferrule System Adapters
For gas chromatography systems
Marca: Trajan™ 0736101
Dettagli aggiuntivi : Peso : 0.20000kg
Descrizione
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifica
Adapter | |
Agilent Capillary Adaptor | |
Agilent Capillary |