Learn More
SGI-1027, TRC
Product Code. 30424196
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424196 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30424196 Supplier Toronto Research Chemicals Supplier No. TRCS28237510MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-[4-[(2-amino-6-methylpyrimidin-4-yl)amino]phenyl]-4-(quinolin-4-ylamino)benzamide
CAS 1020149-73-8
Molecular Formula C27 H23 N7 O
Quantity 10 mg
Synonym N-[4-[(2-Amino-6-methyl-4-pyrimidinyl)amino]phenyl]-4-(4-quinolinylamino)-benzamide
InChI Formula InChI=1S/C27H23N7O/c1-17-16-25(34-27(28)30-17)32-20-10-12-21(13-11-20)33-26(35)18-6-8-19(9-7-18)31-24-14-15-29-23-5-3-2-4-22(23)24/h2-16H,1H3,(H,29,31)(H,33,35)(H3,28,30,32,34)
SMILES Cc1cc(Nc2ccc(NC(=O)c3ccc(Nc4ccnc5ccccc45)cc3)cc2)nc(N)n1
IUPAC Name N-[4-[(2-amino-6-methylpyrimidin-4-yl)amino]phenyl]-4-(quinolin-4-ylamino)benzamide
Molecular Weight (g/mol) 461.518
Formula Weight 461.196