Learn More
Epredia™ Shandon™ Finesse™ ME/E Series Disposable Blade Holders, Low profile
Product Code. 12037159 Shop All Epredia Products
Change view
Click to view available options
Profile:
Low
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Profile Unit Size
12037159 Low Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12037159 Supplier Epredia™ Supplier No. A77510272
  • ONLINE EXCLUSIVE PRICE
    2685.00€
    2505.05€ / pezzi 1
    Risparmia 179.95€
    6% Sconto
In stock
Eliminate the need to switch to a different knife holder for larger specimens by using the Epredia™ Shandon™ Finesse™ ME/E Series Disposable Blade Holders.
  • Improved clamping mechanism design significantly reduces need for knife holder maintenance and improves reliability
  • Consistent reproducible sections provided
  • Larger clamp plate allows easier section retrieval
  • Clamped blade is fully supported along its entire length
  • Entire top stage moves laterally for safer operation
  • Minimized wear on clamping mechanism
TRUSTED_SUSTAINABILITY
Specifications
Description Low-profile
Profile Low
For Use With (Equipment) Standard or Epredia Cassettes