Learn More
Epredia™ Shandon™ Finesse™ Vises and Clamps
Product Code. 12087139 Shop All Epredia Products
Change view
Click to view available options
Product Type:
JB4 Resin Clamp, 0201
Vise, 60 x 55 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12087139 Vise, 60 x 55 mm Each
12017139 JB4 Resin Clamp, 0201 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12087139 Supplier Epredia™ Supplier No. 77510167
  • ONLINE EXCLUSIVE PRICE
    351.00€
    310.16€ / pezzi 1
    Risparmia 40.84€
    11% Sconto
Spedizione stimata: 23-12-2026
Designed for use with Epredia™ Shandon™ and Microm™ Microtomes, the Epredia™ Shandon™ Finesse™ Vises and Clamps meets the needs of most laboratories.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vise, 60 x 55 mm
For Use With (Equipment) Epredia Shandon and Microm Microtomes