Learn More
Epredia™ Peel-A-Way™ Disposable Embedding Molds
Codice prodotto. 12647736 Sfoglia Tutto Epredia Prodotti
Cambia vista
Click to view available options
Dimensioni (L x P x A):
22 x 22 x 20 mm
22 x 30 x 20 mm
22x22 to 12x12 mm, 20 mm deep
Forma:
Rectangular
Square
Truncated, tapered
Dimensione della confezione:
Pezzo
pezzi 288
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensioni (L x P x A) Forma unitSize
12647736 22x22 to 12x12 mm, 20 mm deep Truncated, tapered pezzi 288
12677736 22 x 30 x 20 mm Rectangular pezzi 288
12731548 22 x 22 x 20 mm Square pezzi 288
11919671 22x22 to 12x12 mm, 20 mm deep Truncated, tapered Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 12647736 Fornitore Epredia™ N. del fornitore 1220
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Promote rapid paraffin hardening with Epredia Peel-A-Way Disposable Embedding Molds.
  • Constructed of rigid transparent plastic.
TRUSTED_SUSTAINABILITY
Specifica
Materiale Plastica
Dimensioni (L x P x A) 22x22 to 12x12 mm, 20 mm deep
Forma Truncated, tapered