Learn More
Epredia™ Stainless-Steel Embedding Base Molds
Product Code. 12607916 Shop All Epredia Products
Change view
Click to view available options
Dimensions (L x W x H):
24 x 24 x 5 mm
24 x 30 x 9 mm
24 x 37 x 9 mm
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
12607916 24 x 37 x 9 mm Pack of 10
11306820 24 x 24 x 5 mm Pack of 10
12697906 24 x 30 x 9 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12607916 Supplier Epredia™ Supplier No. 6401018
  • ONLINE EXCLUSIVE PRICE
    143.00€
    110.46€ / pezzi 10
    Risparmia 32.54€
    22% Sconto
 Restricted Item
In stock
Optimize tissue embedding with the superior thermal exchange of our high-quality, durable Stainless-Steel Embedding Base Molds.
  • Sizes fit most cassettes
  • Superior thermal exchange
  • Rounded corners
TRUSTED_SUSTAINABILITY
Specifications
Product Type Base Mold
Material Stainless Steel
Dimensions (L x W x H) 24 x 37 x 9 mm