Learn More
Epredia™ Stainless-Steel Embedding Base Molds
Codice prodotto. 12687906 Sfoglia Tutto Epredia Prodotti
Cambia vista
Click to view available options
Dimensioni (L x P x A):
10 x 10 x 5 mm
24 x 24 x 5 mm
24 x 30 x 9 mm
24 x 37 x 9 mm
Dimensione della confezione:
pezzi 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensioni (L x P x A) unitSize
12687906 10 x 10 x 5 mm pezzi 10
11306820 24 x 24 x 5 mm pezzi 10
12697906 24 x 30 x 9 mm pezzi 10
12607916 24 x 37 x 9 mm pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 12687906 Fornitore Epredia™ N. del fornitore 6401015
Please to purchase this item. Need a web account? Register with us today!
Optimize tissue embedding with the superior thermal exchange of our high-quality, durable Stainless-Steel Embedding Base Molds.
  • Sizes fit most cassettes
  • Superior thermal exchange
  • Rounded corners
TRUSTED_SUSTAINABILITY
Specifica
Product Type Base Mold
Materiale Stainless Steel
Dimensioni (L x P x A) 10 x 10 x 5 mm