Learn More
Rifiuto anatomico
For the safe disposal of anatomical and placenta waste. Sharpsguard™ anatomical 2.5 Waste Containers each come with a placenta bag.
Marca: Daniels Healthcare DD912
Dettagli aggiuntivi : Peso : 9.63000kg
Descrizione
- Rigid polypropylene construction and wall thickness
- Offers outstanding protection
- When container is full to the fill line it must be locked and disposed of safely
- All have clear British Standard markings, user instructions and a visible fill line
Specifica
Rifiuto anatomico | |
2.21 L | |
Rosso, Giallo |